Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAGE 1 Antibody (MZ2E/838), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311463H
Description
MAGE 1 Monoclonal antibody specifically detects MAGE 1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
MAGE 1 | |
Monoclonal | |
HRP | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MZ2E/838 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Antigen MZ2-E, Cancer/testis antigen 1.1, cancer/testis antigen family 1, member 1, CT1.1melanoma antigen family A 1, MAGE-1 antigen, MAGE1A, MAGE1melanoma antigen MAGE-1, melanoma antigen family A, 1 (directs expression of antigen MZ2-E), melanoma-associated antigen 1, melanoma-associated antigen MZ2-E, MGC9326 | |
Recombinant human MAGE 1 protein (Uniprot: P43355) | |
0.1 mL | |
Apoptosis, Cancer, Melanoma Cell Markers, Tumor Suppressors | |
4100 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction