Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAGE 1 Antibody (SR1797), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | MAGE 1 |
---|---|
Clone | SR1797 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Flow Cytometry, Immunofluorescence |
Classification | Monoclonal |
Description
MAGE 1 Monoclonal antibody specifically detects MAGE 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
MAGE 1 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Antigen MZ2-E, Cancer/testis antigen 1.1, cancer/testis antigen family 1, member 1, CT1.1melanoma antigen family A 1, MAGE-1 antigen, MAGE1A, MAGE1melanoma antigen MAGE-1, melanoma antigen family A, 1 (directs expression of antigen MZ2-E), melanoma-associated antigen 1, melanoma-associated antigen MZ2-E, MGC9326 | |
A synthesized peptide derived from human MAGE 1 (Uniprot #: P43355) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1797 | |
Western Blot, Flow Cytometry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Melanoma Cell Markers, Tumor Suppressors | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
4100 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title