Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAGE 1 Rabbit anti-Human, Mouse, Clone: 9Y6K1, Novus Biologicals™

Rabbit Monoclonal Antibody
$207.86 - $463.77
Specifications
| Antigen | MAGE 1 |
|---|---|
| Clone | 9Y6K1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
MAGE 1 Monoclonal antibody specifically detects MAGE 1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| MAGE 1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Melanoma Cell Markers, Tumor Suppressors | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 4100 | |
| IgG | |
| Affinity purified |
| 9Y6K1 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Antigen MZ2-E, Cancer/testis antigen 1.1, cancer/testis antigen family 1, member 1, CT1.1melanoma antigen family A 1, MAGE-1 antigen, MAGE1A, MAGE1melanoma antigen MAGE-1, melanoma antigen family A, 1 (directs expression of antigen MZ2-E), melanoma-associated antigen 1, melanoma-associated antigen MZ2-E, MGC9326 | |
| A synthetic peptide corresponding to a sequence within amino acids 200-309 of human MAGE 1 (P43355). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title