Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAGE1 Monoclonal Antibody (MA454), Abbomax

Catalog No. 502204406
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4406 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4406 Supplier Abbomax Inc Supplier No. 606020
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: breast cancers Cellular location: sytoplasmic.

MAGE1 Monoclonal antibody specifically detects MAGE1 in Human, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen MAGE1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone MA454
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene MAGEA1
Gene Accession No. P43355
Gene Alias antigen MZ2-E; antigen p97 (melanoma associated) identified by monoclonal antibodies 133.2 and 96.5; cancer/testis antigen 1.1; cancer/testis antigen family 1, member 1; CD228; CT1.1; mage 1; MAGE family member A1; MAGE1; mage-1; MAGE-1 antigen; MAGE1A; MAGEA1; Mage-a1; MAP97; melanoma antigen family A 1; melanoma antigen family A, 1; melanoma antigen family A, 1 (directs expression of antigen MZ2-E); melanoma antigen family A1; melanoma antigen MAGE-1; melanoma antigen, family A, 1; melanoma-associated antigen 1; melanoma-associated antigen MZ2-E; Melanoma-associated antigen p97; melanoma-associated antigen p97, isoform 2; melanotransferrin; MELTF; membrane-bound transferrin-like protein; Mfi2; MTf; MTF1
Host Species Mouse
Immunogen A recombinant protein MAGE1
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4100
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.