Learn More
Invitrogen™ MAGE1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117018
Description
Positive Samples: 22Rv1 Immunogen sequence: MSLEQRSLHC KPEEALEAQQ EALGLVCVQA ATSSSSPLVL GTLEEVPTAG STDPPQSPQG ASAFPTTINF TRQRQPSEGS SSREEEGPST SCILESLFRA VITKKVADLV GFLLLKYRAR EPVTKAEMLE SVIKNYKHCF PEIFGKASES LQLVFGIDVK EADPTGHSYV LVTCLGLSYD GLLGDNQIMP KTGFLIIVLV MIAMEGGHAP EEEIWEELSV MEVYDGREHS AYGEPRKLLT QDLVQEKYLE YRQVPDSDPA RYEFLWGPRA LAETSYVKVL EYVIKVSARV RFFFPSLREA ALREEEEGV.
Human malignant neoplasms carry rejection antigens that are recognized by the patients' autologous, tumor directed and specific, cytolytic, CD8+ T lymphocyte clones (CTL). An important group of antigens is coded by the MAGE family of genes. It was identified that melanoma associated with antigens (MAAs). Because MAGE-1 is expressed on a significant proportion of human neoplasms of various hisological tyoes (melanoma, brain tumors of glial origin, neuroblastoma, non-small cell lung cancer, breast, gastric, colorectal, ovarian, renal cell carcinomas) and not on normal tissues, the encoded antigen may serve as a marker of early detection and target for cancer immunotherapy.
Specifications
MAGE1 | |
Polyclonal | |
Unconjugated | |
MAGEA1 | |
antigen MZ2-E; antigen p97 (melanoma associated) identified by monoclonal antibodies 133.2 and 96.5; cancer/testis antigen 1.1; cancer/testis antigen family 1, member 1; CD228; CT1.1; mage 1; MAGE family member A1; MAGE1; mage-1; MAGE-1 antigen; MAGE1A; MAGEA1; Mage-a1; MAP97; melanoma antigen family A 1; melanoma antigen family A, 1; melanoma antigen family A, 1 (directs expression of antigen MZ2-E); melanoma antigen family A1; melanoma antigen MAGE-1; melanoma antigen, family A, 1; melanoma-associated antigen 1; melanoma-associated antigen MZ2-E; Melanoma-associated antigen p97; melanoma-associated antigen p97, isoform 2; melanotransferrin; MELTF; membrane-bound transferrin-like protein; Mfi2; MTf; MTF1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
4100 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P43355 | |
MAGEA1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-309 of human MAGE-1/MAGE1A (NP_0049793). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.