Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGE1 Recombinant Rabbit Monoclonal Antibody (9Y6K1)
GREENER_CHOICE

Catalog No. PIMA537821
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537821 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537821 Supplier Invitrogen™ Supplier No. MA537821
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: HeLa, A-549, A375, Mouse lung, Mouse testis Immunogen sequence: VMIAMEGGHA PEEEIWEELS VMEVYDGREH SAYGEPRKLL TQDLVQEKYL EYRQVPDSDP ARYEFLWGPR ALAETSYVKV LEYVIKVSAR VRFFFPSLRE AALREEEEGV.

Human malignant neoplasms carry rejection antigens that are recognized by the patients' autologous, tumor directed and specific, cytolytic, CD8+ T lymphocyte clones (CTL). An important group of antigens is coded by the MAGE family of genes. It was identified that melanoma associated with antigens (MAAs). Because MAGE-1 is expressed on a significant proportion of human neoplasms of various hisological tyoes (melanoma, brain tumors of glial origin, neuroblastoma, non-small cell lung cancer, breast, gastric, colorectal, ovarian, renal cell carcinomas) and not on normal tissues, the encoded antigen may serve as a marker of early detection and target for cancer immunotherapy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGE1
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 9Y6K1
Concentration 0.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MAGEA1
Gene Accession No. P43355
Gene Alias antigen MZ2-E; antigen p97 (melanoma associated) identified by monoclonal antibodies 133.2 and 96.5; cancer/testis antigen 1.1; cancer/testis antigen family 1, member 1; CD228; CT1.1; mage 1; MAGE family member A1; MAGE1; mage-1; MAGE-1 antigen; MAGE1A; MAGEA1; Mage-a1; MAP97; melanoma antigen family A 1; melanoma antigen family A, 1; melanoma antigen family A, 1 (directs expression of antigen MZ2-E); melanoma antigen family A1; melanoma antigen MAGE-1; melanoma antigen, family A, 1; melanoma-associated antigen 1; melanoma-associated antigen MZ2-E; Melanoma-associated antigen p97; melanoma-associated antigen p97, isoform 2; melanotransferrin; MELTF; membrane-bound transferrin-like protein; Mfi2; MTf; MTF1
Gene Symbols MAGEA1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 200-309 of human MAGE-1/MAGE1A (P43355).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17137, 4100
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.