Learn More
Invitrogen™ MAGEA10 Polyclonal Antibody

Description
The antibody detects endogenous levels of total MAGEA10 protein.
Specifications
Specifications
| Antigen | MAGEA10 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | MAGEA10 |
| Gene Accession No. | P43363 |
| Gene Alias | cancer/testis antigen 1.10; cancer/testis antigen family 1, member 10; CT1.10; MAGE family member A10; MAGE10; MAGE-10 antigen; MAGEA10; melanoma antigen family A, 10; melanoma antigen family A10; melanoma-associated antigen 10; RGD1563693 |
| Gene Symbols | MAGEA10 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.