Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAGEA12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570459
Description
This target displays homology in the following species: Human: 100%.
Not known, though may play a role tumor transformation or progression. In vitro promotes cell viability in melanoma cell lines.
Specifications
MAGEA12 | |
Polyclonal | |
Unconjugated | |
MAGEA12 | |
Cancer/testis antigen 1.12; cancer/testis antigen family 1, member 12; CT1.12; MAGE family member A12; MAGE12; MAGE-12 antigen; MAGE12F antigen; MAGEA12; melanoma antigen family A, 12; melanoma antigen family A12; melanoma-associated antigen 12 | |
Rabbit | |
Affinity chromatography | |
RUO | |
4111 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P43365 | |
MAGEA12 | |
synthetic peptide directed towards the middle region of human MAGEA12 (aa 69-118). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction