Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAGEA12 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987524
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7524 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7524 Supplier Bioss Supplier No. BS6818R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Not known, though may play a role tumor transformation or progression. In vitro promotes cell viability in melanoma cell lines.

Specifications

Antigen MAGEA12
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MAGEA12
Gene Accession No. P43365
Gene Alias Cancer/testis antigen 1.12; cancer/testis antigen family 1, member 12; CT1.12; MAGE family member A12; MAGE12; MAGE-12 antigen; MAGE12F antigen; MAGEA12; melanoma antigen family A, 12; melanoma antigen family A12; Melanoma-associated antigen 12
Gene Symbols MAGEA12
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MAGEA12.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4111
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.