Learn More
Invitrogen™ MAGEA2 Monoclonal Antibody (1B1)

Description
Peptide Sequence: MPLEQRSQHC KPEEGLEARG EALGLVGAQA PATEEQQTAS SSSTLVEVTL GEVPAADSPS PPHSPQGASS FSTTINYTLW RQSDEGSSNQ EEEGPRMFPD LESEFQAAIS RKMVELVHFL LLKYQAREPV TKAEMLESVL RNCQDFFPVI FSKASEYLQL VFGIEVVEVV PISHLYILVT CLGLSYDGLL GDNQVMPKTG LLIIVLAIIA IEGDCAPEEK IWEELSMLEV FEGREDSVFA HPRKLLMQDL VQENYLEYRQ VPGSDPACYE FLWGPRALIE TSYVKVLHHT LKIGGEPHIS YPPLHERALR EGEE.
Specifications
Specifications
| Antigen | MAGEA2 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1B1 |
| Concentration | 0.43 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | MAGEA2 |
| Gene Accession No. | P43356 |
| Gene Alias | Cancer/testis antigen 1.2; cancer/testis antigen family 1, member 2; CT1.2; MAGE family member A2; MAGE2; MAGE-2 antigen; MAGEA2; MAGEA2A; MAGEA2B; melanoma antigen 2; melanoma antigen family A, 2; melanoma antigen family A2; melanoma-associated antigen 2; MGC131923 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.