Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGEA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA513151
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA513151 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA513151 Supplier Invitrogen™ Supplier No. PA513151
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MAGEA2 Polyclonal Antibody for Western Blot, Flow

MAGEA2 is a member of the MAGEA gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 80% sequence identity between each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are expressed at a high level in a number of tumors of various histologic types, and are silent in normal tissues with the exception of testis and placenta. The MAGEA genes are clustered on chromosome Xq28. They may be implicated in some hereditary disorders, such as dyskeratosis congenita. This gene has two identical copies at different loci.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEA2
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene MAGEA2
Gene Accession No. P43356
Gene Alias Cancer/testis antigen 1.2; cancer/testis antigen family 1, member 2; CT1.2; MAGE family member A2; MAGE2; MAGE-2 antigen; MAGEA2; MAGEA2A; MAGEA2B; melanoma antigen 2; melanoma antigen family A, 2; melanoma antigen family A2; melanoma-associated antigen 2; MGC131923
Gene Symbols MAGEA2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 81-108 amino acids from the N-terminal region of human MAGEA2.
Purification Method Ammonium Sulfate Precipitation, Antigen Affinity Chromatography, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4101
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.