Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAGEA3 Antibody (OTI1G9), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272570H
Description
MAGEA3 Monoclonal specifically detects MAGEA3 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MAGEA3 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
MAGEA3 | |
Full length recombinant protein of human MAGEA3 (NP_005353) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI1G9 | |
Western Blot, Immunohistochemistry | |
Antigen MZ2-D, Cancer/testis antigen 1.3, CT1.3 MAGE-3 antigen, HIP8, MAGE3 MAGEA6, melanoma antigen family A, 3, melanoma-associated antigen 3, member 3, MGC14613 | |
Mouse | |
Affinity Purified | |
Cancer | |
4102.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction