Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGEA3 Monoclonal Antibody (OTI1H1)
GREENER_CHOICE

Catalog No. PIMA526488
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526488 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526488 Supplier Invitrogen™ Supplier No. MA526488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MAGEA3 Monoclonal Antibody for Western Blot, IHC (P)

This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEA3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1H1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MAGEA3
Gene Accession No. P43357
Gene Alias Antigen MZ2-D; cancer/testis antigen 1.3; cancer/testis antigen family 1, member 3; CT1.3; HIP8; HYPD; MAGE family member A3; MAGE3; MAGE-3 antigen; MAGEA3; Mage-a3; MAGEA6; melanoma antigen family A, 3; melanoma antigen family A3; melanoma antigen, family A, 3; melanoma-associated antigen 3; MGC14613
Gene Symbols MAGEA3
Host Species Mouse
Immunogen Full length human recombinant protein of MAGEA3 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4102
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.