Learn More
Invitrogen™ MAGEA3 Monoclonal Antibody (OTI2B1)

Description
MAGEA3 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | MAGEA3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI2B1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | MAGEA3 |
| Gene Accession No. | P43357 |
| Gene Alias | Antigen MZ2-D; cancer/testis antigen 1.3; cancer/testis antigen family 1, member 3; CT1.3; HIP8; HYPD; MAGE family member A3; MAGE3; MAGE-3 antigen; MAGEA3; Mage-a3; MAGEA6; melanoma antigen family A, 3; melanoma antigen family A3; melanoma antigen, family A, 3; melanoma-associated antigen 3; MGC14613 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.