Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGEA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597948
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597948 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597948 Supplier Invitrogen™ Supplier No. PA597948
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MAGEA3 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEA3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene MAGEA3
Gene Accession No. P43357
Gene Alias Antigen MZ2-D; cancer/testis antigen 1.3; cancer/testis antigen family 1, member 3; CT1.3; HIP8; HYPD; MAGE family member A3; MAGE3; MAGE-3 antigen; MAGEA3; Mage-a3; MAGEA6; melanoma antigen family A, 3; melanoma antigen family A3; melanoma antigen, family A, 3; melanoma-associated antigen 3; MGC14613
Gene Symbols MAGEA3
Host Species Rabbit
Immunogen Recombinant Human Melanoma-associated antigen 3 protein (2-104AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4102
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.