Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAGEA4 Antibody (CPTC-MAGEA4-1) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279861
Description
MAGEA4 Monoclonal specifically detects MAGEA4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
| MAGEA4 | |
| Monoclonal | |
| 1 mg/mL | |
| Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| MAGE-X2 antigen, melanoma antigen family A, 4, melanoma-associated antigen 4, member 4 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2c κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| CPTC-MAGEA4-1 | |
| Unconjugated | |
| 10 mM PBS with No Preservative | |
| MAGEA4 | |
| Recombinant full-length human MAGEA4 protein (Uniprot: P43358) | |
| 100 μg | |
| Cancer | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction