Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAGEA4 Antibody (CPTC-MAGEA4-1), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279861B
Description
MAGEA4 Monoclonal specifically detects MAGEA4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MAGEA4 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
MAGEA4 | |
Recombinant full-length human MAGEA4 protein (Uniprot: P43358) | |
0.1 mL | |
Cancer | |
Human | |
IgG2c κ |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
CPTC-MAGEA4-1 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
MAGE-X2 antigen, melanoma antigen family A, 4, melanoma-associated antigen 4, member 4 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction