Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAGEA4 Antibody (CPTC-MAGEA4-1), DyLight 680, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP279861FR

Catalog No. NB011285


Only null left
Add to Cart

Description

Description

MAGEA4 Monoclonal specifically detects MAGEA4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

MAGEA4
Monoclonal
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
MAGEA4
Recombinant full-length human MAGEA4 protein (Uniprot: P43358)
0.1 mL
Cancer
Human
IgG2c κ
Immunohistochemistry, Immunohistochemistry (Paraffin)
CPTC-MAGEA4-1
Immunohistochemistry, Immunohistochemistry-Paraffin
MAGE-X2 antigen, melanoma antigen family A, 4, melanoma-associated antigen 4, member 4
Mouse
Protein A or G purified
RUO
Primary
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.