Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGEA4 Monoclonal Antibody (OTI1F9)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA526118

Catalog No. PIMA526118


Only null left
Add to Cart

Description

Description

MAGEA4 Monoclonal Antibody for Western Blot, IHC (P), Flow

MAGEA4 is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MAGEA4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P43358
MAGEA4
Full length human recombinant protein of MAGEA4 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2a
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI1F9
Unconjugated
MAGEA4
Cancer/testis antigen 1.4; cancer/testis antigen family 1, member 4; CT1.4; MAGE family member A4; MAGE4; MAGE-4 antigen; MAGE-41; MAGE-41 antigen; MAGE4A; MAGE4B; MAGEA4; Mage-a4; MAGE-X2; MAGE-X2 antigen; melanoma antigen family A, 4; melanoma antigen family A4; melanoma antigen, family A, 4; melanoma-associated antigen 4; MGC21336
Mouse
Protein A/G
RUO
4103
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.