Learn More
MAGEA4 Mouse anti-Human, Clone: OTI5E8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505423

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita. At least four variants encoding the same protein have been found for this gene.Specifications
MAGEA4 | |
Monoclonal | |
Unconjugated | |
MAGEA4 | |
CT1.4, MAGE-41, MAGE-X2, MAGE4, MAGE4A, MAGE4B | |
Mouse | |
Affinity Chromatography | |
RUO | |
4103 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI5E8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P43358 | |
MAGEA4 | |
Full length human recombit protein of human MAGEA4 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.