Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAGEA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102282
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102282 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102282 Supplier Thermo Scientific Supplier No. PA5102282
Add to Cart
Add to Cart

MAGEA5 Polyclonal Antibody

MAGEA5 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEA5
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MAGEA5
Gene Accession No. P43359
Gene Alias Cancer/testis antigen 1.5; cancer/testis antigen family 1, member 5; CT1.5; MAGE family member A5; MAGE5; MAGE-5 antigen; MAGE-5a antigen; MAGE-5b antigen; MAGEA4; Magea5; Mage-a5; melanoma antigen family A, 5; melanoma antigen family A5; melanoma antigen, family A, 5; melanoma-associated antigen 5; MGC129526
Gene Symbols MAGEA5
Host Species Rabbit
Immunogen A synthesized peptide derived from human MAGEA5(Accession P43359), corresponding to amino acid residues D76-Y124.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17141, 4104
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.