Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAGEA5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987525
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7525 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7525 Supplier Bioss Supplier No. BS6820R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita. This MAGEA gene encodes a protein that is C-terminally truncated compared to other family members, and this gene can be alternatively interpreted to be a pseudogene. The protein is represented in this Gene record in accordance with the assumed protein-coding status defined in the literature. Read-through transcription exists between this gene and the upstream melanoma antigen family A, 10 (MAGEA10) gene. [provided by RefSeq, Oct 2011].

Specifications

Antigen MAGEA5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MAGEA5
Gene Accession No. P43359
Gene Alias Cancer/testis antigen 1.5; cancer/testis antigen family 1, member 5; CT1.5; MAGE family member A5; MAGE5; MAGE-5 antigen; MAGE-5a antigen; MAGE-5b antigen; MAGEA4; MAGEA5; Mage-a5; melanoma antigen family A, 5; melanoma antigen family A5; melanoma antigen, family A, 5; Melanoma-associated antigen 5; MGC129526
Gene Symbols MAGEA5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MAGEA5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4104
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.