Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAGEA6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575647
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
MAGEA6 is a member of the MAGEA gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 80% sequence identity between each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are expressed at a high level in a number of tumors of various histologic types, and are silent in normal tissues with the exception of testis and placenta. The MAGEA genes are clustered on chromosome Xq28. They may be implicated in some hereditary disorders, such as dyskeratosis congenita.
Specifications
MAGEA6 | |
Polyclonal | |
Unconjugated | |
MAGEA6 | |
Cancer/testis antigen 1.6; cancer/testis antigen family 1, member 6; CT1.6; MAGE family member A6; MAGE3B; MAGE-3b; MAGE3B antigen; MAGE6; MAGE-6 antigen; MAGEA6; Mage-a6; melanoma antigen family A, 6; melanoma antigen family A6; melanoma antigen, family A, 6; melanoma-associated antigen 6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17142, 4105 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P43360 | |
MAGEA6 | |
Synthetic peptide corresponding to residues in Human MAGEA6. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction