Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGEA8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552040
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552040 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552040 Supplier Invitrogen™ Supplier No. PA552040
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AQGEAPGLMD VQIPTAEEQK AASSSSTLIM GTLEEVTDSG SPSPPQSPEG ASSSLTVTDS TLWSQSDEGS SSNEEEGPST SPDPAHLESL FREALDEKVA ELVRFLLRKY QIKEPVTKAE MLESVI Highest antigen sequence identity to the following orthologs: Mouse - 35%, Rat - 38%.

This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita. Multiple alternatively spliced variants, encoding the same protein, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEA8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAGEA8
Gene Accession No. P43361
Gene Alias Cancer/testis antigen 1.8; cancer/testis antigen family 1, member 8; CT1.8; mage 8; MAGE family member A8; MAGE8; MAGE-8 antigen; MAGEA8; Mage-a8; melanoma antigen family A, 8; melanoma antigen family A8; melanoma antigen, family A, 8; melanoma-associated antigen 8; MGC2182
Gene Symbols MAGEA8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAGEA8. Recombinant protein control fragment (Product #RP-92336).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4107
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.