Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAGEA9 Antibody (OTI1D8), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272572B
Description
MAGEA9 Monoclonal specifically detects MAGEA9 in Human samples. It is validated for Western Blot.Specifications
MAGEA9 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
MAGEA9 | |
Full length recombinant protein of human MAGEA9 (NP_005356) produced in HEK293T cells. | |
0.1 mL | |
4108.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI1D8 | |
Western Blot | |
Cancer/testis antigen 1.9, CT1.9MAGEA9B, MAGE-9 antigen, MAGE9cancer/testis antigen family 1, member 9, MAGEA9A, melanoma antigen family A, 9, melanoma-associated antigen 9, MGC8421 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction