Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAGEA9 Antibody (OTI1D8), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272572PCP
Description
MAGEA9 Monoclonal antibody specifically detects MAGEA9 in Human samples. It is validated for Western BlotSpecifications
MAGEA9 | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
4108 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot | |
OTI1D8 | |
Western Blot | |
Cancer/testis antigen 1.9, CT1.9MAGEA9B, MAGE-9 antigen, MAGE9cancer/testis antigen family 1, member 9, MAGEA9A, melanoma antigen family A, 9, melanoma-associated antigen 9, MGC8421 | |
Full length human recombinant protein of human MAGEA9 (NP_005356) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction