Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAGEB1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $610.00
Specifications
Antigen | MAGEB1 |
---|---|
Dilution | Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
MAGEB1 Polyclonal specifically detects MAGEB1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MAGEB1 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
4112 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LKVAHATAAEKEECPSSSPVLGDTPTSSPAAGIPQKPQGAPPTTTAAAAVSCTESDEGAKCQGEENASFSQATTSTESSVKDPVAWEAGMLMHFILRKYKMREPIMKADMLKVVDEKYKDHFTEILNGASR | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
Polyclonal | |
Rabbit | |
Human | |
Cancer/testis antigen 3.1, CT3.1cancer/testis antigen family 3, member 1, DAM10MAGE-B1 antigen, DSS/AHC critical interval MAGE superfamily 10, DSS-AHC critical interval MAGE superfamily 10, MAGEL1MAGE-XP antigen, MAGE-Xp, MAGEXP, melanoma antigen family B, 1, melanoma-associated antigen B1, MGC9322 | |
MAGEB1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title