Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGEB1 Monoclonal Antibody (OTI2B12)
GREENER_CHOICE

Catalog No. PIMA526452
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526452 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526452 Supplier Invitrogen™ Supplier No. MA526452
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MAGEB1 Monoclonal Antibody for Western Blot, IHC (P)

This gene is a member of the MAGEB gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 68% sequence identity to each other. The promoters and first exons of the MAGEB genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. This gene is localized in the DSS (dosage-sensitive sex reversal) critical region, and expressed in testis and in a significant fraction of tumors of various histological types. This gene and other MAGEB members are clustered on chromosome Xp22-p21. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene, however, the full length nature of some variants has not been defined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEB1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2B12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MAGEB1
Gene Accession No. P43366
Gene Alias cancer/testis antigen 3.1; cancer/testis antigen family 3, member 1; CT3.1; dam1; DAM10; DSS/AHC critical interval MAGE superfamily 10; DSS-AHC critical interval MAGE superfamily 10; LOC491803; MAGE family member B1; MAGEB1; Mage-b1; MAGE-B1 antigen; Magel1; Mage-rs1; MAGEXP; MAGE-Xp; MAGE-XP antigen; melanoma antigen family B, 1; melanoma antigen family B1; melanoma antigen, family B, 1; melanoma-associated antigen B1; MGC9322; RGD1565746; Smage1
Gene Symbols MAGEB1
Host Species Mouse
Immunogen Full length human recombinant protein of MAGEB1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4112
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.