Learn More
Invitrogen™ MAGEB1 Monoclonal Antibody (OTI4E12)

Description
MAGEB1 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | MAGEB1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI4E12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | MAGEB1 |
| Gene Accession No. | P43366 |
| Gene Alias | cancer/testis antigen 3.1; cancer/testis antigen family 3, member 1; CT3.1; dam1; DAM10; DSS/AHC critical interval MAGE superfamily 10; DSS-AHC critical interval MAGE superfamily 10; LOC491803; MAGE family member B1; MAGEB1; Mage-b1; MAGE-B1 antigen; Magel1; Mage-rs1; MAGEXP; MAGE-Xp; MAGE-XP antigen; melanoma antigen family B, 1; melanoma antigen family B1; melanoma antigen, family B, 1; melanoma-associated antigen B1; MGC9322; RGD1565746; Smage1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.