Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAGEB1 Antibody (OTI4E12), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272573UV
Description
MAGEB1 Monoclonal specifically detects MAGEB1 in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MAGEB1 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MAGEB1 | |
Full length recombinant protein of human MAGEB1 (NP_796379) produced in E.coli. | |
0.1 mL | |
4112.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI4E12 | |
Western Blot, Immunohistochemistry | |
Cancer/testis antigen 3.1, CT3.1cancer/testis antigen family 3, member 1, DAM10MAGE-B1 antigen, DSS/AHC critical interval MAGE superfamily 10, DSS-AHC critical interval MAGE superfamily 10, MAGEL1MAGE-XP antigen, MAGE-Xp, MAGEXP, melanoma antigen family B, 1, melanoma-associated antigen B1, MGC9322 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Canine | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction