Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGEB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551532
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551532 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551532 Supplier Invitrogen™ Supplier No. PA551532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FMNVLGAYDG EEHLIYGEPR KFITQDLVQE KYLKYEQVPN SDPPRYQFLW GPRAYAETTK MKVLEFLAKM NGATPRDFPS HYEEALRDEE ERAQVRSSVR ARRRTTATTF RARSRAPFSR SS Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 53%.

This gene is a member of the MAGEB gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 68% sequence identity to each other. The promoters and first exons of the MAGEB genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. This gene is localized in the DSS (dosage-sensitive sex reversal) critical region, and expressed in testis and in a significant fraction of tumors of various histological types. This gene and other MAGEB members are clustered on chromosome Xp22-p21. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene, however, the full length nature of some variants has not been defined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEB1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAGEB1
Gene Accession No. P43366
Gene Alias cancer/testis antigen 3.1; cancer/testis antigen family 3, member 1; CT3.1; dam1; DAM10; DSS/AHC critical interval MAGE superfamily 10; DSS-AHC critical interval MAGE superfamily 10; LOC491803; MAGE family member B1; MAGEB1; Mage-b1; MAGE-B1 antigen; Magel1; Mage-rs1; MAGEXP; MAGE-Xp; MAGE-XP antigen; melanoma antigen family B, 1; melanoma antigen family B1; melanoma antigen, family B, 1; melanoma-associated antigen B1; MGC9322; RGD1565746; Smage1
Gene Symbols MAGEB1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAGEB1. Recombinant protein control fragment (Product #RP-91938).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4112
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.