Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAGEB10 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306050MFV450
Description
MAGEB10 Polyclonal antibody specifically detects MAGEB10 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| MAGEB10 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| FLJ32965, MAGE family testis and tumor-specific protein, MAGE-B10 antigen, melanoma antigen family B, 10, melanoma-associated antigen B10, MGC120394 | |
| Produced in rabbits immunized with E. coli-derived Human MAGEB10 fragment. | |
| 0.1 mL | |
| Cancer | |
| 139422 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction