Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGEB10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551426
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551426 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551426 Supplier Invitrogen™ Supplier No. PA551426
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LDILEEEEES PPSASACLKD VFQSSLDGAS NNPHGLREAQ STSTSATAAS HTRHPEGVND QMEERPICTQ DLEATDSFPR GPVDEKVIIL VHYLLYKYQM KEPITKADML R Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 30%.

This gene encodes a member of the B subfamily of the melanoma associated antigen protein family. The encoded protein is specifically expressed in testis and tumor cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEB10
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAGEB10
Gene Accession No. Q96LZ2
Gene Alias MAGE family member B10; MAGE family testis and tumor-specific protein; MAGEB10; MAGE-B10 antigen; melanoma antigen family B, 10; melanoma antigen family B10; melanoma-associated antigen B10
Gene Symbols MAGEB10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAGEB10. Recombinant protein control fragment (Product #RP-91972).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139422
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.