Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGEB10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546529
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546529 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546529 Supplier Invitrogen™ Supplier No. PA546529
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MPRGQKSKLR AREKRRQARG GLEDLIDALD ILEEEEESPP SASACLKDVF.

This gene encodes a member of the B subfamily of the melanoma associated antigen protein family. The encoded protein is specifically expressed in testis and tumor cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEB10
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MAGEB10
Gene Accession No. Q96LZ2
Gene Alias MAGE family member B10; MAGE family testis and tumor-specific protein; MAGEB10; MAGE-B10 antigen; melanoma antigen family B, 10; melanoma antigen family B10; melanoma-associated antigen B10
Gene Symbols MAGEB10
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human MAGEB10 (aa 1-50).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139422
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.