Learn More
Invitrogen™ MAGEB16 Polyclonal Antibody

Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: SEDTSDPRNV PADALDQKVA FLVNFMLHKC QMK Highest antigen sequence indentity to the following orthologs: Rat - 48%, Mouse - 48%.
Specifications
Specifications
| Antigen | MAGEB16 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MAGEB16 |
| Gene Accession No. | A2A368 |
| Gene Alias | MAGE family member B16; MAGEB16; MAGE-B16 antigen; melanoma antigen family B, 16 (pseudogene); melanoma antigen family B16; melanoma-associated antigen B16 |
| Gene Symbols | MAGEB16 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.