Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGEB17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552001
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552001 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552001 Supplier Invitrogen™ Supplier No. PA552001
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LYLKEMDPSR QSYVLVGKLD FPNQGSLSDG GGFPLSGLLM VLLSTIFMHG NRATEEEMWE CLNALGMYKG RKHFIYGEPQ ELVTKDLVRE GYLEYQQVPS SDPPRYEFLW GPRARAETSK MKV Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 64%.

MAGEB17 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEB17
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAGEB17
Gene Accession No. A8MXT2
Gene Alias MAGE family member B17; MAGEB17; melanoma antigen family B, 17 (pseudogene); melanoma antigen family B17; melanoma-associated antigen B17
Gene Symbols MAGEB17
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAGEB17. Recombinant protein control fragment (Product #RP-92256).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 645864
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.