Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAGEB18 Antibody (OTI1F5), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272575F
Description
MAGEB18 Monoclonal specifically detects MAGEB18 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
MAGEB18 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
MAGEB18 | |
Full length recombinant protein of human MAGEB18 (NP_775970) produced in HEK293T cells. | |
0.1 mL | |
286514.0 | |
Store at 4°C in the dark. |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI1F5 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
MAGE-B18 antigen, melanoma antigen family B, 18, melanoma-associated antigen B18, MGC33889 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction