Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAGEB2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$254.42
Specifications
| Antigen | MAGEB2 |
|---|---|
| Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
MAGEB2 Polyclonal antibody specifically detects MAGEB2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| MAGEB2 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Cancer/testis antigen 3.2, cancer/testis antigen family 3, member 2, CT3.2MAGE-B2 antigen, DAM6MAGE XP-2 antigen, DSS/AHC critical interval MAGE superfamily 6, DSS-AHC critical interval MAGE superfamily 6, MAGE-XP-2, melanoma antigen family B, 2, melanoma-associated antigen B2, MGC26438 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: RDETRGLNVPQVTEAEEEEAPCCSSSVSGGAASSSPAAGIP | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol | |
| 4113 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title