Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGEB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584683
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584683 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584683 Supplier Invitrogen™ Supplier No. PA584683
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EGLSVVFGLE LNKVNPNGHT YTFIDKVDLT DEESLLSSWD FP Highest antigen sequence indentity to the following orthologs: Mouse - 43%, Rat - 40%.

MAGEB2 is a member of the MAGEB gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 68% sequence identity to each other. The promoters and first exons of the MAGEB genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. This gene is localized in the DSS (dosage-sensitive sex reversal) critical region. It is expressed in testis and placenta, and in a significant fraction of tumors of various histological types. The MAGEB genes are clustered on chromosome Xp22-p21.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEB2
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAGEB2
Gene Accession No. O15479
Gene Alias Cancer/testis antigen 3.2; cancer/testis antigen family 3, member 2; CT3.2; DAM6; DSS/AHC critical interval MAGE superfamily 6; DSS-AHC critical interval MAGE superfamily 6; MAGE family member B2; MAGE XP-2 antigen; MAGEB2; MAGE-B2 antigen; MAGE-XP-2; melanoma antigen family B, 2; melanoma antigen family B2; melanoma-associated antigen B2; MGC26438
Gene Symbols MAGEB2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAGEB2. Recombinant protein control fragment (Product #RP-107410).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4113
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.