Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAGEB3 Antibody (OTI5H8), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272579B
Description
MAGEB3 Monoclonal specifically detects MAGEB3 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MAGEB3 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
MAGEB3 | |
Full length recombinant protein of human MAGEB3 (NP_002356) produced in E.coli. | |
0.1 mL | |
4114.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI5H8 | |
Western Blot, Immunohistochemistry | |
CT3.5, MAGE-B3 antigen, melanoma antigen family B, 3, melanoma-associated antigen B3, member 5 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction