Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAGEB3 Antibody (OTI5H8), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP272579R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272579R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272579R Supplier Novus Biologicals Supplier No. NBP272579R
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MAGEB3 Monoclonal specifically detects MAGEB3 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen MAGEB3
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5H8
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CT3.5, MAGE-B3 antigen, melanoma antigen family B, 3, melanoma-associated antigen B3, member 5
Gene Symbols MAGEB3
Host Species Mouse
Immunogen Full length recombinant protein of human MAGEB3 (NP_002356) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4114.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.