Learn More
Invitrogen™ MAGEB3 Monoclonal Antibody (OTI5H8)

Description
MAGEB3 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | MAGEB3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI5H8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | MAGEB3 |
| Gene Accession No. | O15480 |
| Gene Alias | cancer/testis antigen family 3, member 5; CT3.5; LOW QUALITY PROTEIN: melanoma-associated antigen B3; MAGE family member B3; MAGEB3; Mage-b3; MAGE-B3 antigen; Mage-ps1; Mage-rs3; melanoma antigen family B, 3; melanoma antigen family B3; melanoma antigen, family B, 3; melanoma-associated antigen B3; RGD1562343; Smage3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.