Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAGEB3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599770
Description
Antibody detects endogenous levels of total MAGB3.
This gene is a MAGE-B subfamily member of the MAGE gene family. MAGE family member proteins direct the expression of tumor antigens recognized on a human melanoma by autologous cytolytic T lymphocytes. There are two known clusters of MAGE genes on chromosome X. The members of the MAGE-A subfamily are located in the Xq28 region, while the members of the MAGE-B subfamily are clustered in the Xp21 region.
Specifications
MAGEB3 | |
Polyclonal | |
Unconjugated | |
MAGEB3 | |
cancer/testis antigen family 3, member 5; CT3.5; LOW QUALITY PROTEIN: melanoma-associated antigen B3; MAGE family member B3; MAGEB3; Mage-b3; MAGE-B3 antigen; Mage-ps1; Mage-rs3; melanoma antigen family B, 3; melanoma antigen family B3; melanoma antigen, family B, 3; melanoma-associated antigen B3; RGD1562343; Smage3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
302746, 4114 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O15480 | |
MAGEB3 | |
A synthesized peptide derived from human MAGEB3(Accession O15480), corresponding to amino acid residues M1-K36. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction