Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAGEB3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987527
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7527 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7527 Supplier Bioss Supplier No. BS6823R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene is a MAGE-B subfamily member of the MAGE gene family. MAGE family member proteins direct the expression of tumor antigens recognized on a human melanoma by autologous cytolytic T lymphocytes. There are two known clusters of MAGE genes on chromosome X. The members of the MAGE-A subfamily are located in the Xq28 region, while the members of the MAGE-B subfamily are clustered in the Xp21 region.

Specifications

Antigen MAGEB3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MAGEB3
Gene Accession No. O15480
Gene Alias cancer/testis antigen family 3, member 5; CT3.5; LOC100353564; LOC102153413; MAGE family member B3; MAGEB3; Mage-b3; MAGE-B3 antigen; MAGE-B3-like; Mage-ps1; Mage-rs3; melanoma antigen family B, 3; melanoma antigen family B3; melanoma antigen, family B, 3; Melanoma-associated antigen B3; melanoma-associated antigen B3-like; RGD1562343; Smage3
Gene Symbols MAGEB3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MAGEB3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4114
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.