Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGEB4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544552
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544552 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544552 Supplier Invitrogen™ Supplier No. PA544552
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KEESPSSSSS VLRDTASSSL AFGIPQEPQR EPPTTSAAAA MSCTGSDKGD Sequence homology: Human: 100%.

This gene is a member of the MAGEB gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 68% sequence identity to each other. The promoters and first exons of the MAGEB genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEB genes are clustered on chromosome Xp22-p21. This gene sequence ends in the first intron of MAGEB1, another family member. This gene is expressed in testis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEB4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MAGEB4
Gene Accession No. O15481
Gene Alias cancer/testis antigen family 3, member 6; CT3.6; MAGE family member B4; MAGEB4; MAGE-B4 antigen; melanoma antigen family B, 4; melanoma antigen family B4; melanoma-associated antigen B4; MGC33144
Gene Symbols MAGEB4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human MAGEB4 (aa 35-84).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4115
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.