Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGEB4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550234
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550234 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550234 Supplier Invitrogen™ Supplier No. PA550234
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total MAGEB4 protein.

This gene is a member of the MAGEB gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 68% sequence identity to each other. The promoters and first exons of the MAGEB genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEB genes are clustered on chromosome Xp22-p21. This gene sequence ends in the first intron of MAGEB1, another family member. This gene is expressed in testis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEB4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene MAGEB4
Gene Accession No. O15481
Gene Alias cancer/testis antigen family 3, member 6; CT3.6; MAGE family member B4; MAGEB4; MAGE-B4 antigen; melanoma antigen family B, 4; melanoma antigen family B4; melanoma-associated antigen B4; MGC33144
Gene Symbols MAGEB4
Host Species Rabbit
Immunogen Full length fusion protein of Human MAGEB4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4115
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.