Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGEB6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559756
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559756 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559756 Supplier Invitrogen™ Supplier No. PA559756
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SVPQESQGAS PTGSPDAGVS GSKYDVAANG QDEKSPSTSH DVSVPQESQG ASPTGSPDAG VSGSKYDVAA EGEDEESVSA SQKAIIFKRL SKDAVKKK Highest antigen sequence identity to the following orthologs: Mouse - 29%, Rat - 30%.

This gene is a member of the MAGEB gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 68% sequence identity to each other. The promoters and first exons of the MAGEB genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. This gene is expressed in testis, and in a significant fraction of tumors of various histological types. The MAGEB genes are clustered on chromosome Xp22-p21.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEB6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAGEB6
Gene Accession No. Q8N7X4
Gene Alias cancer/testis antigen 3.4; cancer/testis antigen family 3, member 4; CT3.4; MAGE family member B6; MAGEB6; MAGE-B6; MAGE-B6 antigen; MAGEB6A; melanoma antigen family B, 6; melanoma antigen family B6; melanoma-associated antigen B6
Gene Symbols MAGEB6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAGEB6. Recombinant protein control fragment (Product #RP-99926).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158809
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.