Learn More
Invitrogen™ MAGEB6 Polyclonal Antibody

Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: SSSSSSRACL GDCRRSSDAS IPQESQGVSP TGSPDAVVSY SKSDVAANGQ DEKSPSTSRD.
Specifications
Specifications
| Antigen | MAGEB6 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MAGEB6 |
| Gene Accession No. | Q8N7X4 |
| Gene Alias | cancer/testis antigen 3.4; cancer/testis antigen family 3, member 4; CT3.4; MAGE family member B6; MAGEB6; MAGE-B6; MAGE-B6 antigen; MAGEB6A; melanoma antigen family B, 6; melanoma antigen family B6; melanoma-associated antigen B6 |
| Gene Symbols | MAGEB6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.