Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGEC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552082
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552082 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552082 Supplier Invitrogen™ Supplier No. PA552082
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QIPQSSPEGD DTQSPLQNSQ SSPEGKDSLS PLEISQSPPE GEDVQSPLQN PASSFFSSAL LSIFQSSPES TQSPFEGFPQ SVLQIPVSAA SSSTLVSIFQ Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 31%.

MAGEC1 is a member of the melanoma antigen gene (MAGE) family. The proteins of this family are tumor-specific antigens that can be recognized by autologous cytolytic T lymphocytes. This protein contains a large number of unique short repetitive sequences in front of the MAGE-homologous sequence, thus is about 800 residues longer than the other MAGE proteins. This gene is expressed in a significant proportion of tumors of various histological types, whereas it is silent in normal tissues, except testis. The function of this gene is unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAGEC1
Gene Accession No. O60732
Gene Alias Cancer/testis antigen 7.1; cancer/testis antigen family 7, member 1; CT7; CT7.1; MAGE family member C1; MAGEC1; MAGE-C1 antigen; melanoma antigen family C, 1; melanoma antigen family C1; melanoma-associated antigen C1
Gene Symbols MAGEC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAGEC1. Recombinant protein control fragment (Product #RP-92216).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9947
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.