Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAGEC2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | MAGEC2 |
---|---|
Concentration | 0.26 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-6175
![]() ![]() |
Proteintech Group Inc
102801AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
MAGE-C2/C3 (Melanoma-associated antigen C2, Hepatocellular carcinoma-associated antigen 587) is a 373 amino acid protein encoded by the human gene MAGEC2. Genes of the MAGE family direct the expression of tumor antigens that are recognized on human melanomas by autologous cytolytic T lymphocytes. Among tumor tissues, MAGE-C2/C3 is frequently expressed in seminomas, melanomas, and bladder transitional cell carcinomas. It is also expressed in a significant fraction of head and neck carcinomas, breast carcinomas, nonsmall-cell lung carcinomas, and sarcomas. In hepatocellular carcinoma, there is a inverse correlation between tumor differentiation and protein expression, i. e. the lower the differentiation, the higher percentage of expression. MAGE-C2/C3 is not expressed in normal tissues, except in germ cells in the seminiferous tubules and in Purkinje cells of the cerebellum. It is strongly expressed in spermatogonia and primary spermatocytes, however, at later stages of maturation, expression gradually decreases to become undetectable in mature spermatids.Specifications
MAGEC2 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q9UBF1 | |
51438 | |
MAGEC2 Fusion Protein Ag0418 | |
Primary | |
-20°C | |
MAGEC2 |
0.26 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Cancer/testis antigen 10, CT10, HCA587, MAGE C2 antigen, MAGE E1 antigen, MAGEC2, MAGEE1, melanoma antigen family C, 2, Melanoma associated antigen C2 | |
MAGEC2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title